Magnesium hydroxide ≥95%, Reagent Grade

310093-25G 310093-2KG 310093-500G
220054-511EA 24.62 USD
220054-511 220054-512 220054-513
Magnesium hydroxide ≥95%, Reagent Grade
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.3261 to 2.3631 g/cm³ (20 °C)
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR