Iron(II,III) oxide

Warning

310069-500G 310069-25G
220054-510EA 532.31 USD
220054-510 220054-509
Iron(II,III) oxide
Iron(II,III) oxide
Formula: Fe₃O₄
Melting Pt: 1560 °C
Density: 5.18 g/cm³ (20 °C)
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 12,04087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR