(±)-1,3-Butanediol, anhydrous ≥99%

Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane

309443-100ML 309443-1L
220054-103EA 17.42 USD
220054-103 220054-104
(±)-1,3-Butanediol, anhydrous ≥99%
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 206…208 °C (1013 hPa)
Melting Pt: –54 °C
Density: 1.004 to 1.006 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR