Hydrazine solution ∼35% N₂H₄ in water, Sigma-Aldrich®

Danger

309400-500G 309400-100G 309400-5G
220054-099EA 136.77 USD
220054-099 220054-098 220054-100
Hydrazine solution ∼35% N₂H₄ in water, Sigma-Aldrich®
Hydrazine
Formula: H₄N₂
MW: 32.05 g/mol
MDL Number: MFCD00011417
CAS Number: 302-01-2
EINECS: 206-114-9
UN: 2030
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR