Sodium hydrogen sulfate, Technical Grade

Danger

307823-500G 307823-2.5KG 307823-20KG
220054-381EA 77.08 USD
220054-381 220053-931 220054-380
Sodium hydrogen sulfate, Technical Grade
Sodium hydrogen sulphate
Formula: HNaO₄S
Melting Pt: 181.85 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003531
CAS Number: 7681-38-1
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 12,08729

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR