Disodium metasilicate

Danger

Synonyms: Sodium metasilicate

307815-1KG 307815-25G
220053-929EA 44.23 USD
220053-929 220053-930
Disodium metasilicate
Disodium metasilicate
Formula: Na₂O₃Si
MW: 122.06 g/mol
Melting Pt: ∼72.2 °C
Density: 1.749 g/cm³ (20 °C)
MDL Number: MFCD00003492
CAS Number: 6834-92-0
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 12,08788

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR