Boron triiodide ≥98%

307629-25G 307629-5G
220054-376EA 1712.31 USD
220054-376 220054-377
Boron triiodide ≥98%
Boron triiodide
Formula: BI₃
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 43 °C
Density: 3.35 g/cm³ (20 °C)
MDL Number: MFCD00036286
CAS Number: 13517-10-7
EINECS: 236-857-4
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR