1,2-Dimethoxyethane ≥99.9% for HPLC, Sigma-Aldrich®

Danger

Synonyms: Dimethyl glycol, Ethylene glycol dimethyl ether, DME

307432-2L 307432-1L 307432-100ML
220053-221EA 124 USD
220053-221 220053-220 220053-219
1,2-Dimethoxyethane ≥99.9% for HPLC, Sigma-Aldrich®
1,2-Dimethoxyethane
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: -69 °C
Density: 1.113 g/cm³ (20 °C)
Flash Pt: -6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II
Merck Index: 13,03251

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR