tert-Butyl methyl ether, anhydrous ≥99.8%

Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

306975-100ML 306975-2L 306975-1L 306975-6X1L
220054-074EA 21.29 USD
220054-074 220054-076 220054-075 220054-077
tert-Butyl methyl ether, anhydrous ≥99.8%
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR