1-Decene ≥97.0%

30650-100ML 30650-500ML
220054-269EA 257.38 USD
220054-269 220054-270
1-Decene ≥97.0%
1-Decene
Formula: C₁₀H₂₀
MW: 140.27 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: -66 °C
Density: 0.7408 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00009577
CAS Number: 872-05-9
EINECS: 212-819-2
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR