Sodium methanesulfonate ≥98%

304506-100G 304506-500G 304506-5G
220054-040EA 77.54 USD
220054-040 220054-041 220054-042
Sodium methanesulfonate ≥98%
Sodium methanesulphonate
Formula: CH₃NaO₃S
MDL Number: MFCD00064389
CAS Number: 2386-57-4
EINECS: 219-203-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR