Petroleum spirit, 35…60 °C, anhydrous

Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

300314-2L 300314-1L 300314-100ML
220053-548EA 50.46 USD
220053-548 220053-547 220053-546
Petroleum spirit, 35…60 °C, anhydrous
Petroleum spirit
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR