Sodium tetrachloroaurate dihydrate ≥99%

Warning

Synonyms: Sodium tetrachloroaurate(III) dihydrate

298174-1G 298174-5G
220053-083EA 249.23 USD
220053-083 220053-084
Sodium tetrachloroaurate dihydrate ≥99%
Sodium tetrachloroaurate dihydrate
Formula: NaAuCl₄·2H₂O
MW: 397.8 g/mol
MDL Number: MFCD00149162
CAS Number: 13874-02-7
EINECS: 239-241-3
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR