Methyl difluoroacetate ≥96%

295914-25G 295914-5G
220053-077EA 103.49 USD
220053-077 220053-078
Methyl difluoroacetate ≥96%
Methyl difluoroacetate
Formula: C₃H₄F₂O₂
MW: 110.06 g/mol
Boiling Pt: 84…85 °C (1013 hPa)
Density: 1.272 g/cm³ (20 °C)
MDL Number: MFCD00039276
CAS Number: 433-53-4
EINECS: 207-089-7
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR