o-Xylene ≥98% for HPLC

Danger

Synonyms: 1,2-Dimethylbenzene

295884-1L 295884-2L 295884-100ML
220053-068EA 43.08 USD
220053-068 220053-069 220053-067
o-Xylene ≥98% for HPLC
o-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 30 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR