Dimethylamine, anhydrous ≥99%

Danger

295280-1KG 295280-400G 295280-100G
220052-955EA 397.38 USD
220052-955 220052-956 220052-954
Dimethylamine, anhydrous ≥99%
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Boiling Pt: 6.9 °C (1013 hPa)
Melting Pt: -92.8 °C
Density: 0.938 g/cm³ (20 °C)
Flash Pt: -55 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1032
ADR: 2,
Merck Index: 12,03278

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR