Potassium formate ≥99%

Synonyms: Formic acid potassium salt

294454-500G 294454-25G 294454-1KG
220052-819EA 48.6 USD
220052-819 220052-818 220052-817
Potassium formate ≥99%
Potassium formate
Formula: CHKO₂
MW: 84.12 g/mol
Melting Pt: 165…168 °C
Density: 1.99 g/cm³ (20 °C)
MDL Number: MFCD00013100
CAS Number: 590-29-4
EINECS: 209-677-9
Merck Index: 12,07795

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR