Butyl benzoate ≥99%

Warning

Synonyms: Benzoic acid butyl ester

293296-1KG 293296-250G
220050-993EA 30.15 USD
220050-993 220050-994
Butyl benzoate ≥99%
Butyl benzoate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 249…251 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009439
CAS Number: 136-60-7
EINECS: 205-252-7
Merck Index: 13,01551

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR