Hexane (mixture of isomers) ≥98.5% suitable for HPLC, Sigma-Aldrich®

Danger

Synonyms: Hexanes, Isohexane

293253-2L 293253-200L 293253-4L 293253-4X2L 293253-50L-P2 293253-4X4L 293253-6X1L 293253-1L 293253-200L-LS
220052-616EA 41.85 USD
220052-616 220052-614 220052-617 220052-618 220052-620 220052-619 220052-621 220052-613 220052-615
Hexane (mixture of isomers) ≥98.5% suitable for HPLC, Sigma-Aldrich®
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR