1-Ethyl-3-methylimidazolium methanesulfonate

29164-100G-F 29164-1KG-F
220051-781EA 317 USD
220051-781 220051-782
1-Ethyl-3-methylimidazolium methanesulfonate
1-Ethyl-3-methylimidazolium methanesulphonate
Formula: C₇H₁₄N₂O₃S
MDL Number: MFCD06798171
CAS Number: 145022-45-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR