4-tert-Octylphenol ≥97%

Danger

Synonyms: 4-tert-Octylphenol, 4-(1,1,3,3-Tetramethylbutyl)phenol

290823-1KG 290823-25G
220052-336EA 93.91 USD
220052-336 220052-337
4-tert-Octylphenol ≥97%
4-tert-Octylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 279…281 °C (1013 hPa)
Melting Pt: 80…81 °C
Density: 0.932 g/cm³ (20 °C)
Flash Pt: 147 °C
MDL Number: MFCD00002368
CAS Number: 140-66-9
EINECS: 205-426-2
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR