Sodium dodecylbenzenesulfonate, Technical Grade

Danger

Synonyms: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate

289957-10KG 289957-25G 289957-1KG 289957-500G
220051-422EA 351.08 USD
220051-422 220051-424 220051-423 220051-425
Sodium dodecylbenzenesulfonate, Technical Grade
Sodium dodecylbenzenesulphonate
Formula: C₁₈H₂₉NaO₃S
MW: 348.48 g/mol
Melting Pt: ? 300 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00011508
CAS Number: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR