Gallium trinitrate hydrate ≥99.9%

289892-25G 289892-5G 289892-100G
220051-416EA 378.46 USD
220051-416 220051-417 220051-415
Gallium trinitrate hydrate ≥99.9%
Gallium trinitrate hydrate
Formula: GaN₃O₉·H₂O
MW: 273.75 g/mol
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR