Ethyl difluoroacetate ≥97%

Synonyms: Difluoroacetic ethyl ester

287628-25G 287628-5G
220051-835EA 58.48 USD
220051-835 220051-836
Ethyl difluoroacetate ≥97%
Ethyl difluoroacetate
Formula: C₄H₆F₂O₂
MW: 124.09 g/mol
Boiling Pt: 98 °C (1013 hPa)
Density: 1.1893 g/cm³ (9.8 °C)
MDL Number: MFCD00013578
CAS Number: 454-31-9
EINECS: 207-223-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR