(R)-(-)-2-Amino-3-methyl-1-butanol ≥98%

Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol

284483-5G 284483-1G
220051-143EA 77.97 USD
220051-143 220051-142
(R)-(-)-2-Amino-3-methyl-1-butanol ≥98%
(R)-(-)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Density: 0.93 g/cm³ (20 °C)
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR