(1R)-(-)-Camphor-10-sulfonic acid

Synonyms: L(-)-Camphor-10-sulphonic acid, (1R)-(-)-2-Oxobornane-10-sulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-CSA

282146-25G 282146-100G
220051-116EA 48.89 USD
220051-116 220051-115
(1R)-(-)-Camphor-10-sulfonic acid
(1R)-(-)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00150753
CAS Number: 35963-20-3
EINECS: 252-817-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR