Bis(2-methoxyethyl)ether, anhydrous ≥99.5%

Danger

Synonyms: 2-Methoxyethyl ether, 1-Methoxy-2-(2-methoxyethoxy)ethane, Diethylene glycol dimethyl ether, Dimethyldigol, Diglyme

281662-1L 281662-100ML 281662-2L
220051-282EA 78.15 USD
220051-282 220051-281 220051-283
Bis(2-methoxyethyl)ether, anhydrous ≥99.5%
Bis(2-methoxyethyl)ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III
Merck Index: 13,03187

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR