5-Methyl-2-nitroanisole ≥99%

Warning

Synonyms: Methyl 6-nitro-m-tolyl ether, 3-Methoxy-4-nitrotoluene, 2-Methoxy-4-methyl-1-nitrobenzene, 1-Methoxy-3-methyl-6-nitrobenzene

281514-25G 281514-5G
220051-251EA 339.23 USD
220051-251 220051-252
5-Methyl-2-nitroanisole ≥99%
5-Methyl-2-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007367
CAS Number: 38512-82-2
EINECS: 253-975-1
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR