Phosphorus(III) bromide 1 M in dichloromethane

276820-100ML 276820-800ML
220050-816EA 70.74 USD
220050-816 220050-817
Phosphorus(III) bromide 1 M in dichloromethane
Phosphorus(III) bromide
Formula: PBr₃
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR