3-Fluorothiophenol ≥95%

Synonyms: 3-Fluorobenzene-1-thiol, 1-Fluoro-3-mercaptobenzene, m-Fluorothiophenol, 3-Fluorobenzenethiol

275387-5G 275387-1G
220050-184EA 34.15 USD
220050-184 220050-183
3-Fluorothiophenol ≥95%
3-Fluorothiophenol
Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 164…166 °C (1013 hPa)
Density: 1.18 g/cm³ (30 °C)
MDL Number: MFCD00040227
CAS Number: 2557-77-9
EINECS: 219-876-2
UN: 3071
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR