Dibutyl ether, anhydrous ≥99.3%

Danger

Synonyms: Di-n-butyl ether

271454-2L 271454-100ML 271454-1L
220050-281EA 48.62 USD
220050-281 220050-279 220050-280
Dibutyl ether, anhydrous ≥99.3%
Dibutyl ether
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.767 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009461
CAS Number: 142-96-1
EINECS: 205-575-3
UN: 1149
ADR: 3,III
Merck Index: 13,01568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR