2-Quinolinol ≥99%

Warning

Synonyms: Quinolin-2(1H)-one, Quinolin-2-ol, Carbostyril, 2-Quinolone, 2-Hydroxyquinoline

270873-5G 270873-1G
220049-460EA 57.08 USD
220049-460 220049-459
2-Quinolinol ≥99%
2-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 199 - 200 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006743
CAS Number: 59-31-4
EINECS: 200-420-6
Merck Index: 13,01835

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR