Benzene ≥99.9% suitable for HPLC, Sigma-Aldrich®

Danger

270709-6X1L 270709-2L 270709-4X4L 270709-100ML
220049-682EA 195.08 USD
220049-682 220049-680 220049-681 220050-131
Benzene ≥99.9% suitable for HPLC, Sigma-Aldrich®
Benzene
Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c0146712ab33142aeeb3843b02381e6bda4028&vid=866487d2506bad13295f27385ab4dfe3176e9bb3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR