Tetraiodomethane ≥97%

Warning

Synonyms: Tetraiodomethane, Carbon Tetraiodide

269220-5G 269220-25G
220049-997EA 78.83 USD
220049-997 220049-996
Tetraiodomethane ≥97%
Tetraiodomethane
Formula: CI₄
MW: 519.63 g/mol
Boiling Pt: 130 - 140 °C (1.5 -rr)
Density: 4.32 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001067
CAS Number: 507-25-5
EINECS: 208-068-5
Merck Index: 13,01828

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR