Lead(II) chloride ≥98%

Danger

268690-1KG 268690-250G 268690-5G
220049-981EA 392.31 USD
220049-981 220049-982 220049-983
Lead(II) chloride ≥98%
Lead(II) chloride
Formula: Cl₂Pb
MW: 278.11 g/mol
Boiling Pt: 950 °C (1013 hPa)
Melting Pt: 496 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 13,05422

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR