Ethyl oleate ≥98%

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

268011-25G 268011-5G
220049-484EA 151.85 USD
220049-484 220049-485
Ethyl oleate ≥98%
Ethyl oleate
Formula: C₂₀H₃₈O₂
MW: 310.52 g/mol
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR