Scandium ≥99.9%

261262-250MG 261262-1G
220049-242EA 338.46 USD
220049-242 220049-241
Scandium ≥99.9%
Scandium
Formula: Sc
MW: 44.96 g/mol
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
UN: 3178
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR