3-Chlorophenylhydrazinium chloride ≥97.0%

Synonyms: 1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride

25970-100G-F 25970-25G-F
220049-174EA 128.85 USD
220049-174 220049-175
3-Chlorophenylhydrazinium chloride ≥97.0%
3-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
MDL Number: MFCD02656655
CAS Number: 2312-23-4
EINECS: 219-005-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR