1,2-Dimethoxyethane, anhydrous ≥99.5%

Danger

Synonyms: Dimethyl glycol, Ethylene glycol dimethyl ether, DME

259527-18L 259527-6X1L 259527-100ML 259527-2L 259527-10L 259527-1L
220049-158EA 336.8 USD
220049-158 220049-161 220049-155 220049-160 220049-156 220049-159
1,2-Dimethoxyethane, anhydrous ≥99.5%
1,2-Dimethoxyethane
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 85 °C (1013 hPa)
Melting Pt: –69 °C
Density: 1.113 g/cm³ (20 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3,II
Merck Index: 13,03251

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR