1-Octadecanol ≥99%

Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol

258768-100G 258768-5G
220049-130EA 91.75 USD
220049-130 220049-131
1-Octadecanol ≥99%
1-Octadecanol
Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 56…57 °C
Density: 0.805 to 0.815 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR