Diisobutylaluminium hydride, Reagent Grade

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

256811-1KG 256811-100G
220048-557EA 667.38 USD
220048-557 220048-556
Diisobutylaluminium hydride, Reagent Grade
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Boiling Pt: 95.5…96.5 °C (0.2 mmHg)
Melting Pt: –80 °C
Density: 0.798 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR