Phosphorus(III) bromide ≥99%

Danger

256536-5G 256536-100G 256536-500G
220048-937EA 86.05 USD
220048-937 220048-935 220048-936
Phosphorus(III) bromide ≥99%
Phosphorus(III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –40 °C
Density: 2.8903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR