Titanium(IV) chloride

Danger

254312-50G 254312-10G
220048-523EA 280 USD
220048-523 220048-522
Titanium(IV) chloride
Titanium(IV) chloride
Formula: Cl₄Ti
Boiling Pt: 136 °C (1013 hPa)
Melting Pt: 136.2 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 1838
ADR: 8,II
Merck Index: 13,09555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR