Hexabutylditin

Danger

Synonyms: Hexa-n-butylditin, Hexabutyldistannane, Bis(tributyltin)

251127-50G 251127-10G
220048-265EA 157.15 USD
220048-265 220048-264
Hexabutylditin
Hexabutylditin
Formula: C₂₄H₅₄Sn₂
MW: 580.11 g/mol
Boiling Pt: 160 °C (0.8 hPa)
Density: 1.162 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009417
CAS Number: 813-19-4
EINECS: 212-383-3
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR