2,6-Di-tert-butyl-4-methylpyridine

Warning

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

249505-25G 249505-1G 249505-5G
220048-081EA 513.69 USD
220048-081 220048-080 220048-082
2,6-Di-tert-butyl-4-methylpyridine
2,6-Di-tert-butyl-4-methylpyridine
Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006305
CAS Number: 38222-83-2
EINECS: 253-834-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR