Calcium oxide ≥98%, Reagent Grade

Danger

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

248568-2.5KG 248568-500G
220048-024EA 236.92 USD
220048-024 220048-025
Calcium oxide ≥98%, Reagent Grade
Calcium oxide
Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR