Titanium(IV) ethanolate, Technical Grade

Synonyms: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate

244759-250G 244759-50G
220045-621EA 481.54 USD
220045-621 220045-622
Titanium(IV) ethanolate, Technical Grade
Titanium(IV) ethanolate
Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Boiling Pt: 150 °C (1013 hPa)
Density: 1.107 g/cm³ (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR