2-Hydroxy-p-toluic acid ≥99%

Warning

Synonyms: 2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid

244503-25G 244503-100G
220047-423EA 28.11 USD
220047-423 220047-422
2-Hydroxy-p-toluic acid ≥99%
2-Hydroxy-p-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00002454
CAS Number: 50-85-1
EINECS: 200-068-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR