Sodium perborate tetrahydrate

Danger

Synonyms: Sodium peroxometaborate tetrahydrate

244120-500G 244120-100G
220047-389EA 62.77 USD
220047-389 220047-388
Sodium perborate tetrahydrate
Sodium perborate tetrahydrate
Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60 °C
Density: 1.73 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR