Titanium(IV) butanolate, Reagent Grade

Danger

Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide

244112-100G 244112-5G 244112-500G 244112-2KG
220046-978EA 73.23 USD
220046-978 220047-387 220047-386 220046-979
Titanium(IV) butanolate, Reagent Grade
Titanium(IV) butanolate
Formula: C₁₆H₃₆O₄Ti
MW: 340.34 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.993 g/cm³ (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR